(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR40822-1G OR40822-250MG OR40822-5G
76583-162EA 303.56 USD
76583-162 76583-160 76583-164
(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester 95%
(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester
Formula: C₁₅H₁₉BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32177102
CAS Number: 2280980-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR