1-Bromo-4-prop-2-ynoxy-benzene 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1-Bromo-4-(prop-2-yn-1-yloxy)benzene
OR40819-10G
OR40819-5G
OR40819-1G
76583-142EA
114
USD
76583-142
76583-144
76583-146
1-Bromo-4-prop-2-ynoxy-benzene 99%
1-Bromo-4-prop-2-ynoxy-benzene
Formula:
C₉H₇BrO MW: 211.055 g/mol Boiling Pt: 144 °C (24 mm Hg) Density: 1.468 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD09946935 CAS Number: 33133-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...