5-Bromo[2,2']bipyridinyl

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-2,2'-bipyridine

OR50952-1G OR50952-250MG OR50952-5G
76586-310EA 74.38 USD
76586-310 76586-308 76586-312
5-Bromo[2,2']bipyridinyl
5-Bromo[2,2']bipyridinyl
Formula: C₁₀H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD08234848
CAS Number: 15862-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR