(7-Bromo-1-benzothiophen-2-yl)methanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (7-Bromobenzo[b]thiophen-2-yl)methanol, 7-Bromo-2-(hydroxymethyl)benzo[b]thiophene

OR43532-100MG OR43532-1G OR43532-250MG
76583-796EA 29.26 USD
76583-796 76583-800 76583-798
(7-Bromo-1-benzothiophen-2-yl)methanol
(7-Bromo-1-benzothiophen-2-yl)methanol
Formula: C₉H₇BrOS
Melting Pt: 77…78 °C
Storage Temperature: Ambient
MDL Number: MFCD12032282
CAS Number: 1171926-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR