5-Bromo-3-methylindole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR7820-25G OR7820-5G OR7820-1G
76588-298EA 710.02 USD
76588-298 76588-296 76588-294
5-Bromo-3-methylindole 98%
5-Bromo-3-methylindole
Formula: C₉H₈BrN
Melting Pt: 76…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038746
CAS Number: 10075-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR