3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR7830-1G OR7830-25G OR7830-5G
76588-312EA 20.32 USD
76588-312 76588-316 76588-314
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid 98%
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid
Formula: C₉H₁₇NO₄
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD03490508
CAS Number: 124072-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR