4,6-Dichloropyrimidine-5-carbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR51918-1G OR51918-25G OR51918-5G
76586-876EA 20.87 USD
76586-876 76586-880 76586-878
4,6-Dichloropyrimidine-5-carbonitrile 97%
4,6-Dichloropyrimidine-5-carbonitrile
Formula: C₅HCl₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD08460393
CAS Number: 5305-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR