2-(3-Aminophenyl)acetic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Aminophenylacetic acid

OR4815-1G OR4815-25G
76585-764EA 82.4 USD
76585-764 76585-762 76585-766
2-(3-Aminophenyl)acetic acid
2-(3-Aminophenyl)acetic acid
Formula: C₈H₉NO₂
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00075058
CAS Number: 14338-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR