1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR47823-1G OR47823-25G OR47823-5G
76585-558EA 20.87 USD
76585-558 76585-562 76585-560
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one
Formula: C₆H₅N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD08234923
CAS Number: 3680-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR