2-(Methylsulfonyl)aniline hydrochloride 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR4779-10G OR4779-1G OR4779-5G
76585-556EA 234.3 USD
76585-556 76585-552 76585-554
2-(Methylsulfonyl)aniline hydrochloride 95+%
2-(Methylsulphonyl)aniline hydrochloride
Formula: C₇H₁₀ClNO₂S
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00216484
CAS Number: 205985-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR