5-Amino-2-chloro-3-methylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-2-chloro-3-picoline, 6-Chloro-5-methylpyridin-3-amine

OR42134-1G OR42134-5G
76583-416EA 20.87 USD
76583-416 76583-418
5-Amino-2-chloro-3-methylpyridine
5-Amino-2-chloro-3-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095087
CAS Number: 38186-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR