5-Bromo-1,7-dimethyl-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-1,7-dimethylindazole

OR43557-1G OR43557-250MG
76583-818EA 88.62 USD
76583-818 76583-816
5-Bromo-1,7-dimethyl-1H-indazole
5-Bromo-1,7-dimethyl-1H-indazole
Formula: C₉H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD11226595
CAS Number: 1092352-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR