5-Nitro-2,4-xylidine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2,4-Dimethyl-5-nitroaniline, 5-Nitro-2,4-xylidine
OR6966-1G
OR6966-25G
OR6966-5G
76587-306EA
20.26
USD
76587-306
76587-310
76587-308
5-Nitro-2,4-xylidine
5-Nitro-2,4-xylidine
Formula:
C₈H₁₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00025199 CAS Number: 2124-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...