4-Carboxyphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid

OR7732-100G OR7732-25G OR7732-5G
76588-184EA 88.62 USD
76588-184 76588-182 76588-180
4-Carboxyphenylboronic acid
4-Carboxyphenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94 g/mol
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151801
CAS Number: 14047-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR