4-Methylumbelliferone
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR7969-100G
OR7969-25G
76588-468EA
54
USD
76588-468
76588-466
4-Methylumbelliferone
4-Methylumbelliferone
Formula:
C₁₀H₈O₃ Melting Pt: 190…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006866 CAS Number: 90-33-5 EINECS: 201-986-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...