5-Amino-1-methyl-1H-pyrazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

OR8822-1G OR8822-25G OR8822-5G
76589-114EA 20.87 USD
76589-114 76589-118 76589-116
5-Amino-1-methyl-1H-pyrazole 97%
5-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
Boiling Pt: 95…96 °C/0.5mmHg
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00068156
CAS Number: 1192-21-8
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR