5-Methoxybenzofuran

Supplier: APOLLO SCIENTIFIC
Warning

OR46210-1G OR46210-5G
76584-950EA 276.04 USD
76584-950 76584-946 76584-948
5-Methoxybenzofuran
5-Methoxybenzofuran
Formula: C₉H₈O₂
MW: 148.16
Boiling Pt: 123 °C/20mmHg
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00079770
CAS Number: 13391-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR