4-Morpholinoaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4896-100G OR4896-25G OR4896-5G
76585-872EA 109 USD
76585-872 76585-870 76585-868
4-Morpholinoaniline 98%
4-Morpholinoaniline
Formula: C₁₀H₁₄N₂O
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00006169
CAS Number: 2524-67-6
EINECS: 219-760-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR