Tetrakis(triphenylphosphine)palladium(0)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Pd(PPh3)4Tetrakis(triphenylphosphine)palladium

OR4225-1G OR4225-25G OR4225-5G
76583-512EA 25.47 USD
76583-512 76583-514 76583-516
Tetrakis(triphenylphosphine)palladium(0)
Tetrakis(triphenylphosphine)palladium (0)
Formula: C₇₂H₆₀P₄Pd
MW: 1155.58 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR