4-Amino-3-iodopyridine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Iodopyridin-4-amine3-Iodo-4-pyridylamine

OR4266-1G OR4266-5G
76583-640EA 13.8 USD
76583-640 76583-642
4-Amino-3-iodopyridine
4-Amino-3-iodopyridine
Formula: C₅H₅IN₂
MW: 220.01 g/mol
Melting Pt: 95…101 °C
Storage Temperature: Ambient
MDL Number: MFCD02663853
CAS Number: 88511-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR