5-Amino-2-methoxypyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR4654-100G OR4654-25G
76585-106EA 55 USD
76585-106 76585-104
5-Amino-2-methoxypyridine 97%
5-Amino-2-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.14
Boiling Pt: 265
Melting Pt: 29…31 °C
Density: 1.575 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006264
CAS Number: 6628-77-9
EINECS: 229-612-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR