1,5-Diphenylcarbazone 95%, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: sym-Diphenylcarbazone, Phenylazoameisensäure-2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazide

OR52237-5G OR52237-1G
76587-084EA 220.42 USD
76587-084 76587-082 76587-080 77422-630
1,5-Diphenylcarbazone 95%, Technical Grade
1,5-Diphenylcarbazone
Formula: C₁₃H₁₂N₄O
MW: 240.26 g/mol
Boiling Pt: 440
Melting Pt: 157
Storage Temperature: Ambient
MDL Number: MFCD00003024
CAS Number: 538-62-5
Merck Index: 12,03380

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR