(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

OR43631-1G OR43631-5G OR43631-25G
76583-910EA 20.32 USD
76583-910 76583-912 77421-482
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR