Benzyl nicotinate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR51903-250G OR51903-50G
76586-868EA 110.02 USD
76586-868 76586-866
Benzyl nicotinate 98%
Benzyl nicotinate
Formula: C₁₃H₁₁NO₂
Boiling Pt: 177 °C/8.25mmHg
Melting Pt: 24 °C
Density: 1.165 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00023584
CAS Number: 94-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR