1-Bromo-2-ethynylbenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR42231-1G OR42231-25G OR42231-5G
76583-496EA 34.64 USD
76583-496 76583-500 76583-498
1-Bromo-2-ethynylbenzene 98%
1-Bromo-2-ethynylbenzene
Formula: C₈H₅Br
Boiling Pt: 92…93 °C/20mmHg
Density: 1.443 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD01863358
CAS Number: 766-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR