3-(Dimethylamino)phenol

Supplier: APOLLO SCIENTIFIC
Warning

OR14323-25G OR14323-1G OR14323-100G
76566-952EA 62.14 USD
76566-952 76566-950 76566-954
3-(Dimethylamino)phenol
3-(Dimethylamino)phenol
Formula: C₈H₁₁NO
Boiling Pt: 267 °C
Melting Pt: 80…84 °C
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002264
CAS Number: 99-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR