D-(+)-Xylose

Supplier: APOLLO SCIENTIFIC
Warning

OR7967-100G OR7967-500G
76588-462EA 22.02 USD
76588-462 76588-464
D-(+)-Xylose
D(+)-Xylose
Formula: C₅H₁₀O₅
Melting Pt: 147…158 °C
Density: 1.525 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
EINECS: 200-400-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR