Benzocaine (ethyl-4-aminobenzoate) 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: AethoformEthyl 4-aminobenzoate

OR52189-100G
76587-050EA 41 USD
76587-050
Benzocaine (ethyl-4-aminobenzoate) 98%
Benzocaine (Ethyl 4-aminobenzoate)
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR