Pyridine-2,6-dicarboxylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5062-100G OR5062-25G OR5062-500G
76586-250EA 31.58 USD
76586-250 76586-248 76586-252
Pyridine-2,6-dicarboxylic acid 98%
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
Melting Pt: 238…239 °C
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR