1,8-Diaminonaphthalene 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1,8-naphthylenediamine, 1,8-Naphthalenediamine, Naphthalene-1,8-diamine
OR70111-100G
OR70111-25G
OR70111-500G
76587-482EA
34.64
USD
76587-482
76587-480
76587-484
1,8-Diaminonaphthalene 99%
1,8-Diaminonaphthalene
Formula:
C₁₀H₁₀N₂ Boiling Pt: 205 °C/12mmHg Melting Pt: 62…64 °C Flash Pt: 195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004033 CAS Number: 479-27-6 EINECS: 207-529-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...