2-Amino-3-iodopyridine 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR7027-25G OR7027-5G
76587-578EA 92.7 USD
76587-578 76587-576
2-Amino-3-iodopyridine 99%
2-Amino-3-iodopyridine
Formula: C₅H₅IN₂
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD01651885
CAS Number: 104830-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR