2-Amino-3,5-dichloropyridine 97+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,5-Dichloro-2-pyridylamine, 3,5-Dichloropyridin-2-amine

OR4268-25G OR4268-5G
76583-646EA 23.34 USD
76583-646 76583-644
2-Amino-3,5-dichloropyridine 97+%
2-Amino-3,5-dichloropyridine
Formula: C₅H₄Cl₂N₂
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00006313
CAS Number: 4214-74-8
EINECS: 224-143-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR