2-Bromo-4-iodopyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR4479-1G OR4479-5G
76584-048EA 20.97 USD
76584-048 76584-050
2-Bromo-4-iodopyridine
2-Bromo-4-iodopyridine
Formula: C₅H₃BrIN
Melting Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD03085768
CAS Number: 100523-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR