3-Iodobenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: m-Iodobenzoic acid

OR4998-100G OR4998-25G OR4998-500G
76586-008EA 74.8 USD
76586-008 76586-006 77422-298
3-Iodobenzoic acid 98%
3-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 337
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00002496
CAS Number: 618-51-9
EINECS: 210-555-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR