1,4-Diiodobenzene

Supplier: APOLLO SCIENTIFIC
Warning

OR4220-100G OR4220-500G
76583-450EA 55.02 USD
76583-450 76583-452
1,4-Diiodobenzene
1,4-Diiodobenzene
Formula: C₆H₄I₂
Boiling Pt: 285 °C
Melting Pt: 131…133 °C
Density: 2.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR