5-Nitrosalicylic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Hydroxy-5-nitrobenzoic acid

OR4992-25G OR4992-500G OR4992-100G
76585-986EA 30.56 USD
76585-986 76585-990 76585-988
5-Nitrosalicylic acid 99%
5-Nitrosalicylic acid
Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00007338
CAS Number: 96-97-9
Merck Index: 12,06730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR