4-Bromoanisole 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR4283-100G OR4283-2.5KG OR4283-500G
76583-682EA 20.32 USD
76583-682 76583-686 76583-684
4-Bromoanisole 99%
4-Bromoanisole
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 223
Melting Pt: 10
Density: 1.494 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00000097
CAS Number: 104-92-7
EINECS: 203-252-1
Merck Index: 13,01412

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR