Eisco Glass Narrow Neck Erlenmeyer Flasks
Supplier: Wards
CH0424K CH0424H FL0701 FL0702 CH0424A FL0708 CH0424I FL0705 CH0424B CH0424C CH0424J CH0424G FL0707 FL0703 CH0424E
470343-756EA 100.6 USD
470343-756 470343-764 470343-852 470344-072 470344-080 470344-392 470344-458 470344-548 470344-914 470345-214 470345-320 470345-598 470345-600 470345-924 470345-994
Eisco Glass Narrow Neck Erlenmeyer Flasks
Flasks Erlenmeyer Flasks
Narrow neck borosilicate glass Erlenmeyer flasks with beaded rims and white graduations.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Narrow neck and conical shape prevents spills
  • Chip and crack resistant, fire polished beaded rims
  • Easy-to-read white enamel graduations and specifications for contrast against dark liquids
Narrow neck Erlenmeyer flasks are made of high quality borosilicate 3.3 glass. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C