Sodium bisulfite 58.5 % SO₂ ACS

Supplier: Ricca Chemical
Danger

Synonyms: Sodium hydrogen sulfiteSodium hydrogen sulphite

RDCS0325-500C2
RCRDCS325500C2CS 73.4 USD
RCRDCS325500C2
Sodium bisulfite 58.5 % SO₂ ACS
Sodium hydrogen sulphite
Formula: HNaO₃S
MW: 104.06 g/mol
Boiling Pt: 315 °C
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7631-90-5

Order Now

Specification Test Results

Assay min 58.5 % SO₂
Heavy Metals (as Pb) max 0.001 %
Chloride (Cl) max 0.02 %
Iron (Fe) max 0.002 %
Insoluble Matter max 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR