Potassium hydroxide 45% (w/w) in aqueous solution FCC

Supplier: Ricca Chemical
Danger

Synonyms: Potash causticPotassium hydrateCaustic potash

R6194000-100A
RCR6194000100AEA 132.02 USD
RCR6194000100A
Potassium hydroxide 45% (w/w) in aqueous solution FCC
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: ~100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 44.0 - 46.0 % (w/w)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR