Sodium hydroxide ≥97.0%, pellets ACS

Supplier: Ricca Chemical
Danger

Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

RDCS0530-1000B1
RCRDCS05301000EA 139.47 USD
RCRDCS05301000
Sodium hydroxide ≥97.0%, pellets ACS
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Assay min 97.0 %
Heavy Metals (as Ag) max 0.002 %
Nitrogen Compounds (as N) max 0.001 %
Phosphate (PO₄) max 0.001 %
Sodium Carbonate (Na₂CO₃) max 1.0 %
Sulfate (SO₄) max 0.003 %
Magnesium (Mg) max 0.002 %
Chloride (Cl) max 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR