Sodium hydroxide 33% (w/w)

Supplier: Ricca Chemical
Danger

Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7275000-20F
RCR7275000-20FEA 446.53 USD
RCR7275000-20F
Sodium hydroxide 33% (w/w)
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 32.7 - 33.3 % (w/w)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR