Abbott™ BinaxNOW™ Legionella Urinary Antigen Card Test Kit and Control, Healthcare Products
Supplier: Healthcare Products
MPC 852-000
MPC 852-010
76553-964EA
1200.05
USD
76553-964
76553-928
Abbott™ BinaxNOW™ Legionella Urinary Antigen Card Test Kit and Control, Healthcare Products
Assays
Immunological Assays
BinaxNOW™ Legionella urinary antigen card is a rapid assay for the qualitative detection of Legionella pneumophila serogroup 1 antigen in urine samples from patients with symptoms of pneumonia.
It is intended to aid in the presumptive diagnosis of Legionnaires’ disease caused by L. pneumophila serogroup 1 in conjunction with culture and other methods.