Abbott™ BinaxNOW™ Malaria Test Kits (US and Global) and Control, Healthcare Products
Supplier: Healthcare Products
MPC 665-010 MPC 665-025 MPC 665-000
76553-954EA 65.89 USD
76553-954 76553-896 76553-918
Abbott™ BinaxNOW™ Malaria Test Kits (US and Global) and Control, Healthcare Products
Assays Immunological Assays
The BinaxNOW™ malaria test is an invitro immunochromatographic assay for the qualitative detection of plasmodium antigens circulating in human venous and capillary EDTA whole blood of individuals with signs and symptoms of malarial infection.
  • Results seen in 15 minutes
  • Enables immediate treatment
  • Simple test using only 1 reagent
  • Requires a small blood sample easily collected by fingerstick
  • No need for skilled microscopist
  • Differentiated results allow diagnosis of potentially lethal P. falciparum infection
  • Easy to read, qualitative results
The test targets the histidine-rich protein II (HRPII) antigen specific to Plasmodium falciparum (P.f.) and a pan-malarial antigen common to all four malaria species capable of infecting humans - P. falciparum, P. vivax (P.v.), P. ovale (P.o.), and P. malariae (P.m.).
Intended to aid in the rapid diagnosis of human malaria infections and in the differential diagnosis of Plasmodium falciparum (P.f.) infections from other less virulent malarial infections. Negative results must be confirmed by thin / thick smear microscopy.