Abbott™ DIGIVAL™ BinaxNOW® Interface, Kits and Accessories for Influenza A&B, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC575-000 MPCARFLUSTART MPCARFLU8PK MPCARFLURR MPCARFLUPRT MPCLFR-003 MPCLFR-124 MPCARFLUBD MPC575-080 MPCMCP1810395 MPCARFLUBDPRT MPCGP15
76553-902EA 3444.31 USD
76553-902 76553-946 76553-914 76553-922 76553-966 76553-938 76553-894 76553-908 76553-888 76553-932 76553-886 76553-974
Abbott™ DIGIVAL™ BinaxNOW® Interface, Kits and Accessories for Influenza A&B, Healthcare Products
Assays Immunological Assays
DIGIVAL™ Interface reads and interprets BinaxNOW® lateral flow tests.
  • OBJECTIVITY - Eliminate operator subjectivity
  • CONNECTIVITY - Combine the control of a connected instrument with the speed of a rapid assay
  • ACCURACY - Reduced potential for error- Data input, Test interpretation and Result reporting
  • QUALITY - Ensure quality controls are tested with our QC lockout feature
  • SPEED - Read and interpret approved rapid assays in seconds
DIGIVAL™ accurately reads and interprets lateral flow tests in just seconds, delivering automated, objective results in any healthcare setting. It is a camera based instrument that will detect the presence of and identify a completed lateral flow assay, analyze the intensity of the test and control line and display the results (positive, negative or invalid) on the color touch screen.