Eisco Rubber Pipette Bulbs
Supplier: EISCO SCIENTIFIC LLC
CH0230B CH0230C CH0230G CH0230D CH0230A
470343-916EA 1.39 USD
470343-916 470345-308 470343-844 470345-298 470345-544
Eisco Rubber Pipette Bulbs
Pipettes Pipette Bulbs
For drawing and expelling liquids from pipettes.
  • Made of pliable yet durable rubber
  • For use with serological, and volumetric pipettes
  • Available in a variety of pipette capacities
Red, pear-shaped bulbs made of quality, heavy-duty rubber. Each bulb has a 5 mm diameter hole for inserting serological or volumetric pipettes.