4-Bromo-3-chloro-2-fluorobenzyl alcohol 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC57076-1G PC57076-250MG PC57076-5G
76561-932EA 103.28 USD
76561-932 76560-708 76562-300
4-Bromo-3-chloro-2-fluorobenzyl alcohol 95%
4-Bromo-3-chloro-2-fluorobenzyl alcohol
Formula: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD28142598
CAS Number: 1702154-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR