2,4,5-Trifluorobenzonitrile 96%

Supplier: APOLLO SCIENTIFIC

PC49703-100G PC49703-25G PC49703-5G
76561-042EA 151.4 USD
76561-042 76559-700 76560-508
2,4,5-Trifluorobenzonitrile 96%
2,4,5-Trifluorobenzonitrile
Formula: C₇H₂F₃N
Storage Temperature: Ambient
MDL Number: MFCD00013289
CAS Number: 98349-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR