3-Bromo-5-fluoroanthranilic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2-Amino-3-bromo-5-fluorobenzoic acid
PC32490-1G
PC32490-5G
76559-772EA
20.77
USD
76559-772
76560-658
3-Bromo-5-fluoroanthranilic acid
3-Bromo-5-fluoroanthranilic acid
Formula:
C₇H₅BrFNO₂ MW: 234.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00662509 CAS Number: 259269-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...