4-Chloro-2-fluoro-3-methoxybenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC302974-10G PC302974-1G PC302974-5G
76563-072EA 282.18 USD
76563-072 76561-842 76562-010
4-Chloro-2-fluoro-3-methoxybenzoic acid 98%
4-Chloro-2-fluoro-3-methoxybenzoic acid
Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD19687204
CAS Number: 1169870-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR